Invitrogen
Manufacturer Code:PA575639
Catalog # PIPA575639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human FAIM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAIM 1 FAIM FAIM1 Fas apoptotic inhibitory molecule 1 Fas apoptotic inhibitory molecule; Fas apoptotic inhibitory molecule 1; rFAIM
Gene Aliases: FAIM; FAIM1
UniProt ID: (Human) Q9NVQ4, (Mouse) Q9WUD8, (Rat) Q8R5H8
Entrez Gene ID: (Human) 55179, (Mouse) 23873, (Rat) 140930
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.