Invitrogen
Manufacturer Code:PA570890
Catalog # PIPA570890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human FAHD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylpyruvase FAHD1, mitochondrial; acylpyruvate hydrolase; C16orf36 YISKL; FAH domain-containing protein 1; Fumarylacetoacetate hydrolase domain-containing protein 1; OAA decarboxylase; Oxaloacetate decarboxylase; YISK like/RJD15; YisK-like protein
Gene Aliases: 1110025H10Rik; AV002085; C16orf36; FAHD1; MNCb-4134; RGD1304560; wu:fj09c02; YISKL; zgc:109929
UniProt ID: (Bovine) Q2HJ98, (Human) Q6P587, (Mouse) Q8R0F8, (Rat) Q6AYQ8
Entrez Gene ID: (Horse) 100067938, (Bovine) 509273, (Human) 81889, (Mouse) 68636, (Rat) 302980, (Zebrafish) 553762
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.