Novus Biologicals
Manufacturer Code:NBP258331
Catalog # NBP258331
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MGIMAASRPLSRFWEWGKNIVCVGRNYADHVREMRSAVLSEPVLFLKPSTAYAPEGSPILMPAYTRNLHHELELGVVMGKRCRAVPEAAAMDYVGGYALCLDMTARD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylpyruvase FAHD1, mitochondrial; acylpyruvate hydrolase; C16orf36 chromosome 16 open reading frame 36 DKFZP566J2046 EC 3.- fumarylacetoacetate hydrolase domain containing 1 fumarylacetoacetate hydrolase domain-containing protein 1 MGC74876 YISK like/RJD15 YISKL YisK-like protein; FAH domain-containing protein 1; Fumarylacetoacetate hydrolase domain-containing protein 1; OAA decarboxylase; Oxaloacetate decarboxylase; YISK like/RJD15; YisK-like protein
Gene Aliases: C16orf36; FAHD1; YISKL
UniProt ID: (Human) Q6P587
Entrez Gene ID: (Human) 81889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.