Invitrogen
Manufacturer Code:PA542050
Catalog # PIPA542050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FAH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-diketonase; FAA; Fumarylacetoacetase; Fumarylacetoacetate hydrolase; fumarylacetoacetate hydrolase (fumarylacetoacetase)
Gene Aliases: FAA; FAH; fb59b12; wu:fb59b12; zgc:55316
UniProt ID: (Human) P16930, (Bovine) A5PKH3, (Rat) P25093, (Mouse) P35505
Entrez Gene ID: (Horse) 100049804, (Human) 2184, (Rabbit) 100350127, (Bovine) 508724, (Dog) 100855836, (Rat) 29383, (Mouse) 14085, (Guinea pig) 100733549, (Zebrafish) 322372
Molecular Function:
hydrolase
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.