Proteintech
Manufacturer Code:149281AP150UL
Catalog # 50-172-9843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FAH Fusion Protein Ag6704 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta diketonase FAA FAH Fumarylacetoacetase Fumarylacetoacetate hydrolase; Beta-diketonase; FAA; Fumarylacetoacetase; Fumarylacetoacetate hydrolase; fumarylacetoacetate hydrolase (fumarylacetoacetase)
Gene Aliases: FAH
UniProt ID: (Human) P16930, (Rat) P25093, (Mouse) P35505
Entrez Gene ID: (Human) 2184, (Rat) 29383, (Mouse) 14085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.