Invitrogen
Manufacturer Code:PA527687
Catalog # PIPA527687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 324 and 551 of Human FAF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fas (TNFRSF6) associated factor 1; FAS-associated factor 1; hFAF1; HFAF1s UBXD12 UBXN3A hFAF1 FAS-associated factor 1 TNFRSF6-associated factor 1 UBX domain protein 3A UBX domain-containing protein 12 UBX domain-containing protein 3A; TNFRSF6-associated factor 1; UBX domain protein 3A; UBX domain-containing protein 12; UBX domain-containing protein 3A
Gene Aliases: AA408698; CGI-03; Dffrx; FAF1; Fam; hFAF1; HFAF1s; UBXD12; UBXN3A
UniProt ID: (Human) Q9UNT3, (Mouse) P54731
Entrez Gene ID: (Human) 11124, (Mouse) 14084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.