Invitrogen
Manufacturer Code:PA571542
Catalog # PIPA571542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 303-332 amino acids from the C-terminal region of human FADS6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fatty acid desaturase 6; Fatty acid desaturase 6 11419- FADS6; fatty acid desaturase domain family, member 6
Gene Aliases: BC044804; FADS6; FP18279; OTTMUSG00000021749
UniProt ID: (Human) Q8N9I5, (Mouse) Q80UG1
Entrez Gene ID: (Human) 283985, (Mouse) 328035
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.