Invitrogen
Manufacturer Code:PA571087
Catalog # PIPA571087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human FADS6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fatty acid desaturase 6; fatty acid desaturase domain family, member 6
Gene Aliases: BC044804; FADS6; FP18279; OTTMUSG00000021749; RGD1311950
UniProt ID: (Bovine) A2VE15, (Human) Q8N9I5, (Mouse) Q80UG1
Entrez Gene ID: (Dog) 609709, (Bovine) 523867, (Pig) 100513229, (Human) 283985, (Rabbit) 100356083, (Horse) 100060278, (Mouse) 328035, (Guinea pig) 100715860, (Rat) 303671
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.