Invitrogen
Manufacturer Code:PA569618
Catalog # PIPA569618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat FADS3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytochrome b5-related protein; Delta(13) desaturase; Delta(13) fatty acid desaturase; delta-9 fatty acid desaturase; delta-9-desaturase; Fatty acid desaturase 3; Fatty acid desaturase 3 11419- Cytochrome b5-related protein FADS3 CYB5RP; linoleoyl-CoA desaturase (delta-9-desaturase)-like 3; putative fatty acid desaturase
Gene Aliases: AI464531; CYB5RP; FADS3; LLCDL3; MNCb-0629
UniProt ID: (Human) Q9Y5Q0, (Bovine) A4IFP3, (Rat) Q8K1P9, (Mouse) Q9JJE7
Entrez Gene ID: (Human) 3995, (Bovine) 515925, (Dog) 476062, (Rabbit) 100352453, (Horse) 100059870, (Rat) 286922, (Mouse) 60527, (Guinea pig) 100723901
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.