Invitrogen
Manufacturer Code:PA525285
Catalog # PIPA525285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 96-122 amino acids from the central region of human FADS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA 6-desaturase; D6D; D6D DES6 FADSD6 LLCDL2 SLL0262 TU13 delta(6) desaturase delta(6) fatty acid desaturase delta-6 desaturase delta-6 fatty acid desaturase delta-6-desaturase linoleoyl-CoA desaturase (delta-6-desaturase)-like 2; delta(6) desaturase; Delta(6) fatty acid desaturase; delta-6 desaturase; delta-6 fatty acid desaturase; delta-6-desaturase; Fatty acid desaturase 2; linoleoyl-CoA desaturase (delta-6-desaturase)-like 2
Gene Aliases: D6D; DES6; FADS2; FADSD6; LLCDL2; SLL0262; TU13
UniProt ID: (Human) O95864
Entrez Gene ID: (Human) 9415
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.