Proteintech
Manufacturer Code:141181AP150UL
Catalog # 50-172-9223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FADS Fusion Protein Ag5267 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAD pyrophosphorylase; FAD pyrophosphorylase FAD synthase FAD1 FADS FLAD1 FMN adenylyltransferase PP591 RP11 307C12.7; FAD synthase; FAD synthase region; FAD synthetase; FAD-synthetase; Fad1, flavin adenine dinucleotide synthetase, homolog; Flavin adenine dinucleotide synthase; flavin adenine dinucleotide synthetase; FMN adenylyltransferase; Molybdenum cofactor biosynthesis protein-like region
Gene Aliases: A930017E24Rik; FAD1; FADS; FLAD1; LSMFLAD; PP591
UniProt ID: (Human) Q8NFF5, (Mouse) Q8R123
Entrez Gene ID: (Human) 80308, (Mouse) 319945, (Rat) 751787
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.