Invitrogen
Manufacturer Code:PA517808
Catalog # PIPA517808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Ser194 of human FADD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fas (TNFRSF6)-associated via death domain; FAS-associated death domain protein; FAS-associated death domain protein Fas-associating death domain-containing protein Fas-associating protein with death domain growth-inhibiting gene 3 protein mediator of receptor induced toxicity mediator of receptor-induced toxicity protein FADD; FAS-associating death domain-containing protein; Fas-associating protein with death domain; Growth-inhibiting gene 3 protein; Mediator of receptor induced toxicity; mediator of receptor-induced toxicity; Protein FADD
Gene Aliases: FADD; GIG3; MORT1
UniProt ID: (Human) Q13158
Entrez Gene ID: (Human) 8772
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.