R&D Systems
Manufacturer Code:AF2938
Catalog # AF2938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human FADD Met1-Ser208 Accession # Q13158 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Fas (TNFRSF6)-associated via death domain; Fas (TNFRSF6)-associated via death domain FAS-associated death domain protein FAS-associating death domain-containing protein GIG3 Growth-inhibiting gene 3 protein Mediator of receptor induced toxicity mediator of receptor-induced toxicity MGC8528 MORT1 MORT1Fas-associating protein with death domain protein FADD; FAS-associated death domain protein; FAS-associating death domain-containing protein; Fas-associating protein with death domain; Growth-inhibiting gene 3 protein; Mediator of receptor induced toxicity; mediator of receptor-induced toxicity; Protein FADD
Gene Aliases: FADD; GIG3; MORT1
UniProt ID: (Human) Q13158
Entrez Gene ID: (Human) 8772
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.