Novus Biologicals
Manufacturer Code:NBP255119
Catalog # NBP255119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EAVVHGLNESEASYLITSVELLESKLKTALLDISCVKHIIYVDNKAINKAEYPEGFEIHSMQSVEELGSNPENLGIPPSRPTPSDMAIVMYTSGSTGRPKGVMMHHSNLIAGMTGQCER |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACS4mental retardation X-linked 68 acyl-CoA synthetase 4 acyl-CoA synthetase long-chain family member 4 EC 6.2.1.3 FACL4long-chain 4 LACS 4 LACS4MRX68 lignoceroyl-CoA synthase Long-chain acyl-CoA synthetase 4 long-chain fatty-acid-Coenzyme A ligase 4 long-chain-fatty-acid--CoA ligase 4 mental retardation X-linked 63 MRX63; acyl-CoA synthetase 4; Arachidonate--CoA ligase; fatty-acid-Coenzyme A ligase, long-chain 4; LACS 4; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; Long-chain-fatty-acid--CoA ligase 4
Gene Aliases: ACS4; ACSL4; FACL4; LACS4; MRX63; MRX68
UniProt ID: (Human) O60488
Entrez Gene ID: (Human) 2182
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.