Invitrogen
Manufacturer Code:MA524047
Catalog # PIMA524047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 324926 |
Immunogen | E. coli-derived recombit mouse FABP9/T-FABP Met1-Val132 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 15 kDa perforatorial protein; Fatty acid-binding protein 9; PERF 15; PERF 15 T-FABP FABP-9 Fatty Acid Binding Protein 9 Testis Fatty Acid-Binding Protein 9 Lipid-Binding Protein PERF PERF15 Testis Lipid Binding Protein Testis Lipid-Binding Protein Testis-Type Fatty Acid-Binding Protein T-FABP TLBP; T-FABP; testis lipid binding protein; Testis lipid-binding protein; Testis-type fatty acid-binding protein; TLBP
Gene Aliases: 1700007P10Rik; Fabp9; Perf; Perf15; Tlbp
UniProt ID: (Mouse) O08716
Entrez Gene ID: (Mouse) 21884
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.