Invitrogen
Manufacturer Code:PA547766
Catalog # PIPA547766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human FABP9 Met1-Val132 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: fatty acid binding protein 9, testis; Fatty acid-binding protein 9; lipid-binding protein; PERF 15 T-FABP FABP-9 Fatty Acid Binding Protein 9 Testis Fatty Acid-Binding Protein 9 Lipid-Binding Protein PERF PERF15 Testis Lipid Binding Protein Testis Lipid-Binding Protein Testis-Type Fatty Acid-Binding Protein T-FABP TLBP; T-FABP; testis fatty acid binding protein; testis lipid binding protein; Testis lipid-binding protein; Testis-type fatty acid-binding protein; TLBP
Gene Aliases: FABP9; PERF; PERF15; T-FABP; TLBP
UniProt ID: (Human) Q0Z7S8
Entrez Gene ID: (Human) 646480
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.