Novus Biologicals
Manufacturer Code:NBP300016JF549
Catalog # NB063962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant FABP7/B-FABP (O15540 Met1-Ala132). |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-FABP; B-FABPDKFZp547J2313 BLBPMRG Brain lipid-binding protein Brain-type fatty acid-binding protein FABPBbrain lipid binding protein fatty acid binding protein 7 brain Fatty acid-binding protein 7 fatty acid-binding protein brain Mammary-derived growth inhibitor related mammary-derived growth inhibitor-related; BLBP; Brain lipid-binding protein; Brain-type fatty acid-binding protein; Fatty acid-binding protein 7; Fatty acid-binding protein, brain; hypothetical protein DKFZp547J2313; Mammary-derived growth inhibitor related; mammary-derived growth inhibitor-related
Gene Aliases: B-FABP; BLBP; FABP7; FABPB; MRG
UniProt ID: (Human) O15540
Entrez Gene ID: (Human) 2173
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.