R&D Systems
Manufacturer Code:AF3166
Catalog # AF3166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human FABP7/B-FABP Met1-Ala132 Accession # O15540 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B-FABP; BFABP B-FABP B-FABPDKFZp547J2313 BLBP BLBPMRG Brain lipid-binding protein Brain-type fatty acid-binding protein FABPBbrain lipid binding protein fatty acid binding protein 7 brain Fatty acid-binding protein 7 fatty acid-binding protein brain Mammary-derived growth inhibitor related mammary-derived growth inhibitor-related; BLBP; Brain lipid-binding protein; Brain-type fatty acid-binding protein; Fatty acid-binding protein 7; Fatty acid-binding protein, brain; hypothetical protein DKFZp547J2313; Mammary-derived growth inhibitor related; mammary-derived growth inhibitor-related
Gene Aliases: B-FABP; BLBP; FABP7; FABPB; MRG
UniProt ID: (Human) O15540
Entrez Gene ID: (Human) 2173
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.