Novus Biologicals
Manufacturer Code:NBP290126H
Catalog # NB070157
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 9 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human FABP6 (AAH22489.1 Met1-Ala128). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: fatty acid binding protein 6 ileal Fatty acid-binding protein 6 gastrotropin I-15PIntestinal 15 kDa protein I-BABPIntestinal bile acid-binding protein I-BALB I-BAP ILBP3 ILBPGT ileal bile acid binding protein ILLBPIleal lipid-binding protein illeal lipid-binding protein; fatty acid binding protein 6, ileal; Fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; ILBP; ileal bile acid binding protein; Ileal lipid-binding protein; illeal lipid-binding protein; Intestinal 15 kDa protein; Intestinal bile acid-binding protein
Gene Aliases: FABP6; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ILLBP
UniProt ID: (Human) P51161
Entrez Gene ID: (Human) 2172
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.