Abnova
Manufacturer Code:H00002172M01
Catalog # 89-022-047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4A4 |
Immunogen | FABP6 (AAH22489 1 a.a. ~ 128 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FABP6.
For Research Use Only
Protein Aliases: fatty acid binding protein 6, ileal; Fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ileal bile acid binding protein; Ileal lipid-binding protein; ILLBP; illeal lipid-binding protein; Intestinal 15 kDa protein; Intestinal bile acid-binding protein
Gene Aliases: FABP6; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ILLBP
UniProt ID: (Human) P51161
Entrez Gene ID: (Human) 2172
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.