R&D Systems
Manufacturer Code:MAB1678
Catalog # MAB1678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 242305 |
Immunogen | <EM>E. coli</EM>-derived recombinant human FABP3/H-FABP Met1-Ala133 Accession # P05413 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: FABP11 fatty acid binding protein 11 fatty acid binding protein 3 muscle and heart (mammary-derived growthinhibitor) Fatty acid-binding protein 3 Fatty acid-binding protein 3 muscle fatty acid-binding protein heart Heart-type fatty acid-binding protein HFABP H-FABP H-FABPM-FABP Mammary-derived growth inhibitor MDGI Muscle fatty acid-binding protein O-FABP; fatty acid binding protein 11; fatty acid binding protein 3, muscle and heart; Fatty acid-binding protein 3; Fatty acid-binding protein, heart; H-FABP; Heart-type fatty acid-binding protein; M-FABP; Mammary-derived growth inhibitor; MDGI; Muscle fatty acid-binding protein
Gene Aliases: FABP11; FABP3; H-FABP; M-FABP; MDGI; O-FABP
UniProt ID: (Human) P05413
Entrez Gene ID: (Human) 2170
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.