Invitrogen
Manufacturer Code:PA595317
Catalog # PIPA595317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human Cardiac FABP identical to the related mouse and rat sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fatty acid binding protein 11; Fatty acid binding protein 3 heart; fatty acid binding protein 3, muscle and heart; fatty acid binding protein heart 1; fatty acid binding protein heart 4; Fatty acid-binding protein 3; Fatty acid-binding protein heart Fatty acid-binding protein 3 Heart-type fatty acid-binding protein H-FABP Mammary-derived growth inhibitor MDGI Muscle fatty acid-binding protein M-FABP FABP3 FABP11 MDGI; Fatty acid-binding protein, heart; H-FABP; heart fatty acid binding protein; Heart-type fatty acid-binding protein; M-FABP; Mammary-derived growth inhibitor; MDGI; Muscle fatty acid-binding protein
Gene Aliases: FABP11; FABP3; Fabph-1; Fabph-4; Fabph1; Fabph4; H-FABP; M-FABP; MDGI; O-FABP
UniProt ID: (Human) P05413, (Mouse) P11404, (Rat) P07483
Entrez Gene ID: (Human) 2170, (Mouse) 14077, (Rat) 79131
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.