Invitrogen
Manufacturer Code:PA528957
Catalog # PIPA528957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 133 of Human MDGI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FABP11 H-FABP M-FABP MDGI O-FABP Fatty acid-binding protein 3 muscle fatty acid binding protein 11 fatty acid-binding protein heart heart-type fatty acid-binding protein mammary-derived growth inhibitor muscle fatty acid-binding protein; fatty acid binding protein heart 1; fatty acid binding protein heart 4; Fatty acid-binding protein 3; Fatty acid-binding protein, heart; H-FABP; Heart-type fatty acid-binding protein; Mammary-derived growth inhibitor; MDGI
Gene Aliases: Fabp3; Fabph-1; Fabph-4; Fabph1; Fabph4; H-FABP; Mdgi
UniProt ID: (Mouse) P11404
Entrez Gene ID: (Mouse) 14077
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.