Abnova
Manufacturer Code:PAB19591
Catalog # 89-121-119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human FABP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of FABP3.
For Research Use Only
Protein Aliases: FABP11; fatty acid binding protein 11; fatty acid binding protein 3, muscle and heart; Fatty acid-binding protein 3; Fatty acid-binding protein, heart; H-FABP; Heart-type fatty acid-binding protein; M-FABP; Mammary-derived growth inhibitor; MDGI; Muscle fatty acid-binding protein; O-FABP
Gene Aliases: FABP11; FABP3; H-FABP; M-FABP; MDGI; O-FABP
UniProt ID: (Human) P05413
Entrez Gene ID: (Human) 2170
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.