Proteintech
Manufacturer Code:212521AP150UL
Catalog # 50-173-3338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FABP2 Fusion Protein Ag15793 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FABP2 FABPI Fatty acid binding protein 2 I FABP; FABPI; fatty acid binding protein 1; fatty acid binding protein 2, intestinal; Fatty acid-binding protein 2; Fatty acid-binding protein, intestinal; I-FABP; intestinal fatty acid binding protein; intestinal fatty acid binding protein 2; Intestinal-type fatty acid-binding protein
Gene Aliases: FABP; FABP2; FABPI; I-FABP
UniProt ID: (Human) Q2NKJ1, (Mouse) P55050, (Rat) P02693
Entrez Gene ID: (Human) 2169, (Mouse) 14079, (Rat) 25598
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.