Invitrogen
Manufacturer Code:PA576451
Catalog # PIPA576451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human FABP12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FABP12 {ECO:0000303|PubMed:18786628}; fatty acid binding protein ORF; Fatty acid-binding protein 12; fatty acid-binding protein 12 {ECO:0000303|PubMed:18786628}; intracellular fatty acid-binding protein FABP12; Probable fatty acid-binding protein ENSP00000353650; probable fatty acid-binding protein ENSP00000353650 homolog
Gene Aliases: 1700008G05Rik; FABP12; RGD1565000
UniProt ID: (Human) A6NFH5, (Rat) B7SUM8, (Mouse) Q9DAK4
Entrez Gene ID: (Human) 646486, (Rat) 499570, (Mouse) 75497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.