R&D Systems
Manufacturer Code:MAB29644SP
Catalog # MAB29644SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 965616 |
Immunogen | <em>E. coli</em>-derived recombinant human FABP1/L-FABP Met1-Ile127 Accession # P07148 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
Protein Aliases: FABPL fatty acid binding protein 1 liver fatty acid-binding protein liver LFABP L-FABP L-FABPFatty acid-binding protein 1 Liver-type fatty acid-binding protein; fatty acid binding protein 1, liver; Fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein
Gene Aliases: FABP1; FABPL; L-FABP
UniProt ID: (Human) P07148
Entrez Gene ID: (Human) 2168
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.