Invitrogen
Manufacturer Code:MA535080
Catalog # PIMA535080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0545 |
Immunogen | A synthesized peptide derived from human FABP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 14 kDa selenium-binding protein; 14 kDa selenium-binding protein FABP1 Fabpl Fabplg Fatty acid-binding protein 1 Fatty acid-binding protein liver L-FABP Liver-type fatty acid-binding protein p14 SCP SCP. Squalene- and sterol-carrier protein Z-protein; fatty acid binding protein 1, liver; fatty acid binding protein liver; Fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein; p14; SCP; Squalene- and sterol-carrier protein; Z-protein
Gene Aliases: FABP1; FABPL; Fabplg; L-FABP; p14; SCP; SCP.
UniProt ID: (Human) P07148, (Mouse) P12710, (Rat) P02692
Entrez Gene ID: (Human) 2168, (Mouse) 14080, (Rat) 24360
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.