Invitrogen
Manufacturer Code:720242
Catalog # PI720242
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptides corresponding to Human FABP1 (aa 70-80 27-40) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14 kDa selenium-binding protein; Fatty Acid Binding Protein 1 FABPL; fatty acid binding protein 1, liver; fatty acid binding protein liver; Fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein; p14; SCP; Squalene- and sterol-carrier protein; Z-protein
Gene Aliases: FABP1; FABPL; Fabplg; L-FABP; p14; SCP; SCP.
UniProt ID: (Human) P07148, (Mouse) P12710, (Rat) P02692
Entrez Gene ID: (Human) 2168, (Mouse) 14080, (Rat) 24360
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.