Abnova
Manufacturer Code:H00002166M07J
Catalog # 89-335-078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H8 |
Immunogen | FAAH (NP_001432 480 a.a. &sim 579 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody.
Protein Aliases: Anandamide amidohydrolase 1; FAAH-1 MGC102823 MGC138146; Fatty acid ester hydrolase; Fatty-acid amide hydrolase 1; Oleamide hydrolase 1
Gene Aliases: FAAH; FAAH-1; FAAH1; PSAB
UniProt ID: (Human) O00519
Entrez Gene ID: (Human) 2166
Molecular Function:
hydrolase
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.