Invitrogen
Manufacturer Code:PA575991
Catalog # PIPA575991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human FAAH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adamide amidohydrolase 1 Adamide amidohydrolase FA2H FAAH FAAH-1 FAAH1 Fatty acid 2-hydroxylase Fatty acid alpha-hydroxylase Fatty acid amide hydrolase Fatty-acid amide hydrolase 1 MGC102823 MGC138146 Oleamide hydrolase 1 Oleamide hydrolase; amide hydrolase; Anandamide amidase; Anandamide amidohydrolase 1; fatty acid amide hydrolase protein; Fatty acid ester hydrolase; Fatty-acid amide hydrolase 1; Oleamide hydrolase 1
Gene Aliases: AW412498; FAAH; FAAH-1; FAAH1; PSAB
UniProt ID: (Human) O00519, (Rat) P97612, (Mouse) O08914
Entrez Gene ID: (Human) 2166, (Rat) 29347, (Mouse) 14073
Molecular Function:
hydrolase
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.