Proteintech
Manufacturer Code:179091AP150UL
Catalog # 50-173-1869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FAAH Fusion Protein Ag12304 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adamide amidohydrolase 1 FAAH FAAH 1 FAAH1 fatty acid amide hydrolase Fatty acid amide hydrolase 1 Oleamide hydrolase 1; amide hydrolase; Anandamide amidase; Anandamide amidohydrolase 1; fatty acid amide hydrolase protein; Fatty acid ester hydrolase; Fatty-acid amide hydrolase 1; Oleamide hydrolase 1
Gene Aliases: AW412498; FAAH; FAAH-1; FAAH1; PSAB
UniProt ID: (Human) O00519, (Rat) P97612, (Mouse) O08914
Entrez Gene ID: (Human) 2166, (Rat) 29347, (Mouse) 14073
Molecular Function:
hydrolase
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.