Invitrogen
Manufacturer Code:PA5100512
Catalog # PIPA5100512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FA2H corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FA2H Faah FAH1 Fatty acid 2-hydroxylase fatty acid alpha-hydroxylase fatty acid hydroxylase domain containing 1 Faxdc1 FLJ25287 G630055L08Rik RGD1310347 SCS7 spastic paraplegia 35 (autosomal recessive) SPG35 WD repeat domain 59 Wdr59; Fatty acid 2-hydroxylase; Fatty acid alpha-hydroxylase; fatty acid hydroxylase domain containing 1; Fatty acid hydroxylase domain-containing protein 1; spastic paraplegia 35 (autosomal recessive); WD repeat domain 59
Gene Aliases: FA2H; FAAH; FAH1; FAXDC1; G630055L08Rik; RGD1310347; SCS7; SPG35; Wdr59
UniProt ID: (Human) Q7L5A8, (Rat) Q2LAM0, (Mouse) Q5MPP0
Entrez Gene ID: (Human) 79152, (Rat) 307855, (Mouse) 338521
Molecular Function:
hydroxylase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.