Proteintech
Manufacturer Code:214811AP150UL
Catalog # 50-173-3441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | F9 Fusion Protein Ag15723 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Christmas factor; Christmas factor coagulation factor IX F9 FIX HEMB PTC; Coagulation factor IX; Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; factor 9; factor IX F9; plasma thromboplastic component; Plasma thromboplastin component; PTC
Gene Aliases: F9; F9 p22; FIX; HEMB; P19; PTC; THPH8
UniProt ID: (Human) P00740
Entrez Gene ID: (Human) 2158
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.