Novus Biologicals
Manufacturer Code:NB600404SS
Catalog # NB600404SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | CI-A3-1 |
Immunogen | Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ADGRE1 Adhesion G protein-coupled receptor E1 cell surface glycoprotein F4/80 DD7A5-7 EGF-like module containing mucin-like hormone receptor-like sequence 1 EGF-like module receptor 1 EGF-like module-containing mucin-like hormone receptor-like 1 EGF-TM7 EMR1 hormone receptor F4/80 Gpf480 Ly71 lymphocyte antigen 71 Mouse EMR1 Rat EMR1 TM7LN3; Adhesion G protein-coupled receptor E1; egf-like module containing, mucin-like, hormone receptor-like 1; egf-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EMR1 hormone receptor
Gene Aliases: ADGRE1; EMR1; TM7LN3
UniProt ID: (Human) Q14246
Entrez Gene ID: (Human) 2015
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.