Abnova
Manufacturer Code:PAB27699
Catalog # 89-129-215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acid at N-terminus of human F2R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of F2R.
For Research Use Only
Protein Aliases: CF2R; coagulation factor II (thrombin) receptor; Coagulation factor II receptor; HTR; PAR-1; PAR1; protease activated receptor 1; protease-activated receptor 1; Proteinase-activated receptor 1; Thrombin receptor; TR
Gene Aliases: CF2R; F2R; HTR; PAR-1; PAR1; TR
UniProt ID: (Human) P25116
Entrez Gene ID: (Human) 2149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.