Abnova
Manufacturer Code:H00002149B02
Catalog # 89-005-328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | F2R (AAH02464.1 1 a.a. ~ 425 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human F2R protein.
For Research Use Only
Protein Aliases: CF2R; coagulation factor II (thrombin) receptor; Coagulation factor II receptor; HTR; PAR-1; PAR1; protease activated receptor 1; protease-activated receptor 1; Proteinase-activated receptor 1; Thrombin receptor; TR
Gene Aliases: CF2R; F2R; HTR; PAR-1; PAR1; TR
UniProt ID: (Human) P25116
Entrez Gene ID: (Human) 2149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.