Invitrogen
Manufacturer Code:PA570162
Catalog # PIPA570162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human F131C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf117 RP11-5P18.9 protein FAM131C; family with sequence similarity 131, member C; Protein FAM131C
Gene Aliases: C1orf117; FAM131C; Gm693; zgc:110264
UniProt ID: (Human) Q96AQ9
Entrez Gene ID: (Human) 348487, (Horse) 100053694, (Rabbit) 100347023, (Dog) 607065, (Guinea pig) 100713015, (Mouse) 277743, (Rat) 690880, (Zebrafish) 619198
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.