Invitrogen
Manufacturer Code:PA557441
Catalog # PIPA557441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human F1142 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 133K02 C5orf3 protein FAM114A2; Protein FAM114A2; testis tissue sperm-binding protein Li 81P
Gene Aliases: 133K02; 1810073G14Rik; 9030624B09Rik; C5orf3; FAM114A2
UniProt ID: (Human) Q9NRY5, (Mouse) Q8VE88
Entrez Gene ID: (Human) 10827, (Mouse) 67726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.