Invitrogen
Manufacturer Code:PA571100
Catalog # PIPA571100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human F102B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family with sequence similarity 102, member B; Protein FAM102B; Protein FAM102B-like protein; sym-3 homolog B
Gene Aliases: 1600010D10Rik; B430201A12Rik; FAM102B; SYM-3B
UniProt ID: (Human) Q5T8I3, (Mouse) Q8BQS4
Entrez Gene ID: (Horse) 100057881, (Human) 284611, (Dog) 490130, (Rabbit) 100341363, (Mouse) 329739, (Guinea pig) 100725800
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.