Novus Biologicals
Manufacturer Code:NB10040810
Catalog # NB10040810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QDYEEKTKKAER corresponding to internal region according to NP_003370.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CVIL CVL Cytovillin cytovillin 2 DKFZp762H157 EZR ezrin FLJ26216 p81 VIL2MGC1584 villin 2 (ezrin) villin-2; Cytovillin; cytovillin 2; epididymis secretory protein Li 105; Ezrin; p81; villin 2 (ezrin); Villin-2
Gene Aliases: CVIL; CVL; EZR; HEL-S-105; VIL2
UniProt ID: (Human) E1P5A8
Entrez Gene ID: (Human) 7430
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.