Proteintech
Manufacturer Code:260561AP150UL
Catalog # 50-173-5186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Ezrin Fusion Protein Ag23530 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CVIL CVL Cytovillin EZR Ezrin p81 VIL2 Villin 2; Cytovillin; cytovillin 2; epididymis secretory protein Li 105; Ezrin; p81; villin 2; villin 2 (ezrin); Villin-2
Gene Aliases: AW146364; CVIL; CVL; EZR; HEL-S-105; p81; R75297; VIL2
UniProt ID: (Human) E1P5A8, (Mouse) P26040
Entrez Gene ID: (Human) 7430, (Mouse) 22350
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.