Novus Biologicals
Manufacturer Code:NBP229948
Catalog # NBP229948
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This EXT2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 182-209 amino acids from the Central region of human EXT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.1.224 EC 2.4.1.225 exostoses (multiple) 2 exostosin 2 exostosin-2 Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase Multiple exostoses protein 2 N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase Putative tumor suppressor protein EXT2 SOTV; Exostosin-2; Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Multiple exostoses protein 2; N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase; Putative tumor suppressor protein EXT2
Gene Aliases: EXT2; SOTV; SSMS
UniProt ID: (Human) Q93063
Entrez Gene ID: (Human) 2132
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.