Novus Biologicals
Manufacturer Code:NBP216395
Catalog # NBP216395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human EXTL3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: botv DKFZp686C2342 EC 2.4.1.223 exostoses (multiple)-like 3 exostosin-like 3 EXTL1L EXTR1BOTV EXT-related protein 1 Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase Hereditary multiple exostoses gene isolog KIAA0519 Multiple exostosis-like protein 3 Putative tumor suppressor protein EXTL3 REG reg receptor REGR RPR; exostoses (multiple)-like 3; exostosin tumor-like 3; Exostosin-like 3; exostosin-like glycosyltransferase 3; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Hereditary multiple exostoses gene isolog; Multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; reg receptor
Gene Aliases: BOTV; EXTL1L; EXTL3; EXTR1; KIAA0519; REGR; RPR
UniProt ID: (Human) O43909
Entrez Gene ID: (Human) 2137
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.