R&D Systems
Manufacturer Code:MAB2536SP
Catalog # MAB2536SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 339509 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 2/EXTL2 Asn43-Met330 Accession # Q9ES89 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-14-N-acetylhexosaminyltransferase EXTL2 alpha-14-N-acteylhexosaminyltransferase Alpha-GalNAcT EXTL2 EC 2.4.1.223 exostoses (multiple)-like 2 Exostosin like 2 exostosin-like 2 EXTL2 EXTR2 EXT-related protein 2 Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase processed exostosin-like 2; Alpha-GalNAcT EXTL2; exostoses (multiple)-like 2; Exostosin-like 2; exostosin-like glycosyltransferase 2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Processed exostosin-like 2
Gene Aliases: EXTL2; EXTR2
UniProt ID: (Human) Q9UBQ6
Entrez Gene ID: (Human) 2135
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.