Novus Biologicals
Manufacturer Code:NBP214953
Catalog # NBP214953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human EXOSC8. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA421P11.3 CBP-interacting protein 3 CIP3 EAP2 exosome complex exonuclease RRP43 exosome component 8Opa-interacting protein 2 OIP2Ribosomal RNA-processing protein 43 Opa interacting protein 2 p9RP11-421P11.3 RRP43OIP-2 Rrp43p; CBP-interacting protein 3; Exosome complex component RRP43; exosome complex exonuclease RRP43; Exosome component 8; OIP-2; Opa interacting protein 2; Opa-interacting protein 2; p9; Ribosomal RNA-processing protein 43
Gene Aliases: bA421P11.3; CIP3; EAP2; EXOSC8; OIP2; p9; PCH1C; RRP43; Rrp43p
UniProt ID: (Human) Q96B26
Entrez Gene ID: (Human) 11340
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.