Novus Biologicals
Manufacturer Code:NBP250173F
Catalog # NBP250173F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | V80P1C10&asteriskC |
Immunogen | Peptide Sequence - KLLMSSTKGLY |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Chronic myelogenous leukemia tumor antigen 28; CML28 exosome complex exonuclease RRP46 exosome component 5Chronic myelogenous leukemia tumor antigen 28 exosome component Rrp46 hRrp46p MGC12901 p12BMGC111224 RRP41B Rrp46p RRP46Ribosomal RNA-processing protein 46; Exosome complex component RRP46; exosome complex exonuclease RRP46; Exosome component 5; exosome component Rrp46; p12B; Ribosomal RNA-processing protein 46
Gene Aliases: CML28; EXOSC5; hRrp46p; p12B; RRP41B; RRP46; Rrp46p
UniProt ID: (Human) Q9NQT4
Entrez Gene ID: (Human) 56915
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.