Novus Biologicals
Manufacturer Code:NBP297516JF549
Catalog # NB044446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human Exosome component 10 fragment. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: autoantigen PM-SCL; autoantigen PM-SCL EC 3.1.13.- exosome component 10 P100 polymyositis-scleroderma overlap syndrome-associated autoantigen p2 p3 p4 PM/Scl-100Autoantigen PM/Scl 2 PM-Scl PMSCL2PMSCL Polymyositis/scleroderma autoantigen 2 polymyositis/scleroderma autoantigen 2 (100kD) polymyositis/scleroderma autoantigen 2 100kDa RRP6 Rrp6p RRP6Polymyositis/scleroderma autoantigen 100 kDa; Autoantigen PM/Scl 2; Exosome component 10; P100 polymyositis-scleroderma overlap syndrome-associated autoantigen; PM/Scl-100; Polymyositis/scleroderma autoantigen 100 kDa; Polymyositis/scleroderma autoantigen 2
Gene Aliases: EXOSC10; p2; p3; p4; PM-Scl; PM/Scl-100; PMSCL; PMSCL2; RRP6; Rrp6p
UniProt ID: (Human) Q01780
Entrez Gene ID: (Human) 5394
Molecular Function:
RNA binding protein
exoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.