Novus Biologicals
Manufacturer Code:NBP222242
Catalog # NBP222242
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 389 and 439 of human Exosome Component 9 using the numbering given in entry NP_005024.2 (GeneID 5393). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
For Research Use Only
Protein Aliases: Autoantigen PM/Scl 1; Autoantigen PM/Scl 1 EC 3.1.13 exosome component 9Polymyositis/scleroderma autoantigen 75 kDa p6 PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen PMSCL175kD Polymyositis/scleroderma autoantigen 1 polymyositis/scleroderma autoantigen 1 (75kD) polymyositis/scleroderma autoantigen 1 75kDa; Exosome complex component RRP45; exosome complex exonuclease RRP45; Exosome component 9; P75 polymyositis-scleroderma overlap syndrome associated autoantigen; P75 polymyositis-scleroderma overlap syndrome-associated autoantigen; PM/Scl-75; PMSCL autoantigen, 75kD; Polymyositis/scleroderma autoantigen 1; polymyositis/scleroderma autoantigen 1, 75kDa; Polymyositis/scleroderma autoantigen 75 kDa
Gene Aliases: EXOSC9; p5; p6; PM/Scl-75; PMSCL1; RRP45; Rrp45p
UniProt ID: (Human) Q06265
Entrez Gene ID: (Human) 5393
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.