Abnova
Manufacturer Code:MAB16123
Catalog # 89-999-007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 14G1 |
Immunogen | Recombinant His tag fusion protein corresponding to rat Exoc4 from <i>E. coli</i>. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ALR; augmenter of liver regeneration (ALR) pseudogene; Exocyst complex component 4; Exocyst complex component Sec8; rSec8; Sec8; SEC8-like 1; Sec8l1; secretory protein SEC8
Gene Aliases: ALR; Exoc4; Sec8; Sec8l1
UniProt ID: (Rat) Q62824
Entrez Gene ID: (Rat) 116654
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.